
5-Chlorosalicylanilide ≥98.0% (by titrimetric analysis)
TCIAC0286-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:207.0 to 211.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:247.68 g/mol
:225-065-4
:GHS07
:4638-48-6
:C₁₃H₁₀ClNO₂
:MFCD00002320