Order Entry
Poland
ContactUsLinkComponent
N6-Dibenzoyladenosine-2',3'-dibenzoate ≥98.0% (by HPLC, titration analysis)
N6-Dibenzoyladenosine-2',3'-dibenzoate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
N6-Dibenzoyladenosine-2',3'-dibenzoate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together