2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone ≥95.0% (by GC)
Catalog # TCIAB1839-25G
Supplier: TCI
CAS Number:
Specifications
- Ilość:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:128.0 to 133.0 deg-C
Specification Test Results
About this item
:
:334.54 g/mol
EINECS:212-990-3
:GHS07
Storage Temperature:Lodówka
CAS Number:903-19-5
Formula:C₂₂H₃₈O₂
MDL Number:MFCD00128797