Ethyl decanoate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-38-3
:
Formula:C₁₂H₂₄O₂
MDL Number:MFCD00009581
:200.32 g/mol
Merck Index:13,03811
Boiling Pt:245 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.862 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-761-9
Specification Test Results
- Solubility:0.016 g/l (20 °C)
- Melting Point:-20 °C
- Molar Mass:200.32 g/mol
- Boiling Point:244 - 246 °C (1013 hPa)
- Vapor Pressure:0.02 hPa (20 °C)
- Flash Point:104 °C
- Refractive Index:1.4248 (20 °C, 589 nm)
- Explosion Limit:0.7 %(V)
- Density:0.86 g/cm3 (20 °C)
- Ignition Point:310 °C
Specification Test Results
Frequently Bought Together