Triacetin (glycerol triacetate), Sigma-Aldrich®
: Merck
:102-76-1
:
:C₉H₁₄O₆
:MFCD00008716
:218.21 g/mol
:13,09664
:258 °C (1013 hPa)
:4 °C
:1.1596 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:138 °C (closed cup)
:Ambient
:203-051-9
Frequently Bought Together