Triacetin (glycerol triacetate) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102-76-1
:
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (760 mmHg)
Melting Pt:3 °C
Density:1.155 g/cm³
Flash Pt:138 °C
Storage Temperature:Ambient
EINECS:203-051-9
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4330 (20°C, 589 nm)
- Density:(25°C) 1.154 to 1.158 g/ml
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together