Order Entry
Poland
ContactUsLinkComponent
Methyl acetoacetate 99+%
Methyl acetoacetate 99+%
Certificates
undefined
Methyl acetoacetate 99+%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4175 to 1.4200 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤20 APHA
  • Free acid (CH3COOH):
    ≤0.2 %

Specification Test Results

Related Information

Frequently Bought Together