

Iron(III) sulphate hydrate 21-23% (Fe basis), Puriss. p.a., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:15244-10-7
:
Formula:Fe₂(SO₄)₃·xH₂O
MDL Number:MFCD00011007
:399.88 g/mol (anhy)
Merck Index:13,04065
Density:3,1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-072-9
UN:3260
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay of Fe:21 - 23 %
- Insoluble in HCl:Max. 0.01 %
- Arsenic (As):Max. 0.0005 %
- Calcium (Ca):Max. 0.01 %
- Copper (Cu):Max. 0.002 %
- Iron (Fe 2⁺):Max. 0.02 %
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.01 %
- Sodium (Na):Max. 0.01 %
- Zinc (Zn):Max. 0.005 %
- Chloride (Cl):Max. 0.005 %
- Nitrate (NO₃):Max. 0.02 %
- Phosphate (PO₄):Max. 0.003 %
Specification Test Results
Frequently Bought Together