(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:76189-55-4
:
Formula:C₄₄H₃₂P₂
MDL Number:MFCD00010805
Merck Index:13,01223
Melting Pt:239…242 °C
:M-Clarity™ quality level = MQ100
:622.69 g/mol
Frequently Bought Together