Order Entry
Poland
ContactUsLinkComponent
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Catalog # MOLEM60347605
Supplier:  Molekula
CAS Number:  
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Catalog # MOLEM60347605
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Merck Index:13,01223
Melting Pt:239…242 °C
 :622.69 g/mol
CAS Number:76189-55-4
Formula:C₄₄H₃₂P₂
MDL Number:MFCD00010805