Order Entry
Poland
ContactUsLinkComponent
(1S)-(-)-Borneol for synthesis, Sigma-Aldrich®
Flammable
(1S)-(-)-Borneol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
(1S)-(-)-Borneol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    205 - 208 °C
  • Molar Mass:
    154.25 g/mol
  • Boiling Point:
    210 °C (1036 hPa)
  • Flash Point:
    66 °C

Specification Test Results

Related Information

Frequently Bought Together