


Boron trifluoride dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13319-75-0
:
Formula:BF₃·2H₂O
MDL Number:MFCD00149901
:103.84 g/mol
Boiling Pt:58.5…60 °C (1 mmHg)
Melting Pt:6.2 °C
Density:1.636 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-569-5
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:2851
:GHS04
:GHS06
:GHS05
Frequently Bought Together