Copper(II) sulphate 0.1 M CuSO4 volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Density:1.02 g/cm³ (20 °C)
EINECS:231-847-6
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
Frequently Bought Together