Order Entry
Poland
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,1,1-Trichloroethane Solution
Supelco® Organic Reference Standard, 1,1,1-Trichloroethane Solution
  40010-U
 :  Merck
 :  
Supelco® Organic Reference Standard, 1,1,1-Trichloroethane Solution
  40010-U
 :  Merck
 :  

 

  • Pk:
    1
  • Standards:
    Certified reference material, 5000 mug/ml in methanol
  • CAS No.:
    71-55-6
  • Description:
    1,1,1-Trichloroethane solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370 - H420
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Methyl chloroform
  • Formula:
    C₂H₃Cl₃
  • Flash point:
    11 °C

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311 - P502