Thallium standard solution, 1000 mg/L Tl in 0,5 M nitric acid (from TlNO₃) Certipur® standard for AAS, Supelco®
: Merck
Thallium standard solution, 1000 mg/L Tl in 0,5 M nitric acid (from TlNO₃) Certipur® standard for AAS, Supelco®
: Merck
: Traceable to SRM from NIST.
- M-Clarity quality level:MQ100
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
BD Microlance™ 3 Needles
: BD Medical
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Single-use syringes, 2-piece, HENKE-JECT®
: HENKE SASS WOLF
Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for AAS, Specpure®
: Thermo Fisher Scientific
IGEPAL® CA-630
: Thermo Fisher Scientific