Order Entry
Poland
ContactUsLinkComponent
VWR® ARISTAR®, Palladium Standard Solution, 10000 mg/l Pd in 10% Hydrochloric Acid (from Pd), Standard for ICP
VWR® ARISTAR®, Palladium Standard Solution, 10000 mg/l Pd in 10% Hydrochloric Acid (from Pd), Standard for ICP
Catalog # 455772X
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Palladium Standard Solution, 10000 mg/l Pd in 10% Hydrochloric Acid (from Pd), Standard for ICP
Catalog # 455772X
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Palladium
  • Symbol:
    Pd

Specifications

About this item

Pd in HCl 10%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.