Sulphur standard solution, 1000 mg/L S in water (from H₂SO₄) Certipur® standard for ICP, Supelco®
numer katalogowy 1.70355.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Sulphur
- Symbol:S
Specifications
About this item
Certifications: Traceable to SRM from NIST.