(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-79-8
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00063238
:132.16 g/mol
Merck Index:14,5213
Boiling Pt:188…190 °C
Melting Pt:–27 °C
Density:1.064
Flash Pt:80 °C (176 °F)
Storage Temperature:Ambient
EINECS:202-888-7
Related Information
Frequently Bought Together