Calcium hypochlorite 65% available chlorine in aqueous solution tech., granular
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7778-54-3
:
Formula:CaCl₂O₂
MDL Number:MFCD00010900
Molecular Weight:142.98 g/mol
Melting Pt:100 °C
Merck Index:13,02111
EINECS:231-908-7
UN:3485
:GHS07
:GHS05
:GHS09
:GHS03
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white to light green Granules or granular solid
- Additional info:typical composition:
- pH:(1% aq. soln.) about 11.6
- Titration with Na2S2O3:≥65 % (Available Cl)
Specification Test Results
Related Information
Frequently Bought Together