Sodium hexametaphosphate ≥96%, crystalline +200 mesh, Sigma-Aldrich®
Catalog # SIAL305553-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:12,08814
Density:2,4 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:272-808-3
CAS Number:68915-31-1
:10124-56-8
Formula:(NaO₃P)n
MDL Number:MFCD00134118