Hydriodic acid 57% in aqueous solution stabilised for analysis, distilled
:10034-85-2
:
:HI
:MFCD00011347
:127 °C (1013 hPa)
:1.7 g/cm³
:Refrigerator
:8,II
:Stabilised with hypophosphorous acid 0.75 %
:127.91 g/mol
:233-109-9
:1787
:GHS05
- Appearance:Clear light yellow to amber Solution (May darken on storage)
- Stabilizer:0.75 % H3PO2
- Titration with NaOH:≥55.0 %
Frequently Bought Together