Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) for qualitative determination of reducing sugars
Catalog # 31964.294
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
Synonyms: Fehling B solution
:
UN:1824
:GHS05
:6381-59-5
Density:1.2 g/cm³ (20 °C)
ADR:8 II