
Ethanol-[D₁] 99.5+ atom % D for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:925-93-9
:
Formula:C₂H₅DO
:47.06 g/mol
Boiling Pt:78 °C (1013 hPa)
Melting Pt:–114.5 °C
Density:0.8 g/cm³
Flash Pt:12 °C
EINECS:213-128-9
UN:1170
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.5 % (Coulometric)
- D-Enrichment:≥99.50 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together