Order Entry
Poland
ContactUsLinkComponent
Gentamicin sulphate 50 mg/ml in aqueous solution, Sigma-Aldrich®
Gentamicin sulphate 50 mg/ml in aqueous solution, Sigma-Aldrich®
Catalog # SIALG1397-10ML
Supplier:  Merck
CAS Number:  
Gentamicin sulphate 50 mg/ml in aqueous solution, Sigma-Aldrich®
Catalog # SIALG1397-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:1405-41-0
MDL Number:MFCD00270181