Order Entry
Poland
ContactUsLinkComponent
Methyl cellulose, 300 - 560 mPa.s, Methocel® A4C
Methyl cellulose, 300 - 560 mPa.s, Methocel® A4C
Catalog # 298944N
Supplier:  VWR Chemicals
CAS Number:  
undefined
Methyl cellulose, 300 - 560 mPa.s, Methocel® A4C
Catalog # 298944N
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Viscosity:
    300 - 560 mPa.s
  • pH (1 %):
    5.0 - 8.0
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Loss on drying:
    Max. 5.0 %

Specification Test Results

About this item

Synonyms: Cellulose methyl ether
 :
Storage Temperature:Ambient
CAS Number:9004-67-5
MDL Number:MFCD00081763