Human Recombinant IL-1B (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 1 beta (IL-1β) is a pro-inflammatory cytokine that is produced by monocytes, macrophages, and dendritic cells (DCs). IL-1β signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter.
- High quality
- Low endotoxin
- Affordable
IL-1β and the independently regulated IL-1α protein have overlapping pro-inflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together