Butyl acetate, Burdick & Jackson™
Catalog # HONC65530-175KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:175 kg
- Seq:4656
Specifications
Specification Test Results
- Assay (GC):Min. 99 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.10 %
- 1-Butanol (GC):Max. 0.10 %
- Free acid (as CH₃COOH):Max. 0.01 %
Specification Test Results
About this item
:
:116.16 g/mol
Boiling Pt:126.2 °C
Density:0.881 g/cm3
Flash Pt:27 °C
UN:1123
ADR:3,III
:GHS07
:GHS02
CAS Number:123-86-4
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445