Order Entry
Poland
ContactUsLinkComponent
Dihexylammonium acetate solution 0.5 M in water, LiChropur™ for ion pair chromatography, Supelco®
Dihexylammonium acetate solution 0.5 M in water, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 93645-25ML
Supplier:  Merck
CAS Number:  
Dihexylammonium acetate solution 0.5 M in water, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 93645-25ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless
  • Appearance (Form):
    Liquid
  • Ttration (Method):
    Non Aqueous Titration
  • Titration (Molarity):
    0.45 - 0.55 mol/l
  • Density D20/4:
    0.992 - 0.996
  • Refractive Index N20/D:
    1.350 - 1.354
  • Test by UV Spectroscopy:
    Corresponds to Requirements
  • Residue (Filter Test):
    No Residue
  • Filtertest (Method):
    Neat
  • Recommended Retest Period:
    42 Months

Specification Test Results

About this item

 :
 :245.41 g/mol
Density:0.992...0.996 g/cm³ (20 °C)
CAS Number:366793-17-1
Formula:C₁₄H₃₁NO₂
MDL Number:MFCD03093619