Order Entry
Poland
ContactUsLinkComponent
Magnesium ionophore III, Selectophore™, function tested ionophore, Supelco®
Magnesium ionophore III, Selectophore™, function tested ionophore, Supelco®
Catalog # 63086-50MG-F
Supplier:  Merck
CAS Number:  
undefined
Magnesium ionophore III, Selectophore™, function tested ionophore, Supelco®
Catalog # 63086-50MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :538.81 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:119110-38-2
Formula:C₃₀H₅₈N₄O₄
MDL Number:MFCD00144715