Lithium ionophore VIII, Selectophore™, function tested ionophore, Supelco®
Catalog # 62571-50MG-F
Supplier: Merck
CAS Number:
Specifications
- Ilość:50 mg
- Seq:2370
Specifications
About this item
:
:798.19 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:133338-85-9
Formula:C₄₈H₈₃N₃O₆
MDL Number:MFCD00467019