
5-Butylpicolinic acid ≥99.0% (by HPLC) derivatisation reagent, for HPLC, Supelco®
Catalog # 55952-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
CAS Number:536-69-6
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD00006298