Order Entry
Poland
ContactUsLinkComponent
Curcumin ≥99.5% (by HPLC) matrix substance for MALDI-MS, Supelco®
Curcumin ≥99.5% (by HPLC) matrix substance for MALDI-MS, Supelco®
Catalog # 78246-100MG
Supplier:  Merck
CAS Number:  
Curcumin ≥99.5% (by HPLC) matrix substance for MALDI-MS, Supelco®
Catalog # 78246-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Color):
    Yellow to Very Dark Yellow and Orange to Very Dark Orange
  • Appearance (Form):
    Powder or Crystals
  • Purity (HPLC area %):
    ≥ 99.5 %
  • Melting Point:
    183 - 188 C
  • Carbon Content:
    68.47 % (theory)
  • Hydrogen Content:
    5.47 % (theory)
  • Proton NMR Spectrum:
    Conforms to structure
  • Metal trace Analysis (ICP):
    Corresponds to requirements
  • BARIUM (ICP):
    ≤ 5 mg/kg
  • CALCIUM (ICP):
    ≤ 100 mg/kg
  • CADMIUM (ICP):
    ≤ 5 mg/kg
  • COBALT (ICP):
    ≤ 5 mg/kg
  • CHROMIUM (ICP):
    ≤ 5 mg/kg
  • COPPER (ICP):
    ≤ 5 mg/kg
  • IRON (ICP):
    ≤ 30 mg/kg
  • POTASSIUM (ICP):
    ≤ 50 mg/kg
  • MAGNESIUM (ICP):
    ≤ 30 mg/kg
  • MANGANESE (ICP):
    ≤ 5 mg/kg
  • SODIUM (ICP):
    ≤ 50 mg/kg
  • NICKEL (ICP):
    ≤ 5 mg/kg
  • LEAD (ICP):
    ≤ 5 mg/kg
  • ZINC (ICP):
    ≤ 5 mg/kg
  • RECOMMENDED RETEST PERIOD:
    36 MONTHS

Specification Test Results

About this item

 :
 :368.39 g/mol
Melting Pt:183…188 °C
Storage Temperature:Refrigerator
CAS Number:458-37-7
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365