Order Entry
Poland
ContactUsLinkComponent
Mercury(II) nitrate monohydrate ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS
  86176.180
 :  
undefined
Mercury(II) nitrate monohydrate ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS
  86176.180
 :  
  • Pk:
    100 g
  • Seq:
    3401
 :
 :342.62 g/mol
 :79 °C
 :4.3 g/cm³ (20 °C)
 :233-152-3
 :1625
 :6.1,II
 :GHS06
 :GHS08
 :GHS09
 :7783-34-8
 :13465-31-1
 :Hg(NO₃)₂·1H₂O
 :MFCD00149736