Order Entry
Poland
ContactUsLinkComponent
 
Hydrogen peroxide solution ≥30%, Semiconductor grade (Honeywell 17948)
Catalog # HONC40267H-5L
CAS Number:  
Hydrogen peroxide solution ≥30%, Semiconductor grade (Honeywell 17948)
Catalog # HONC40267H-5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay:
    Min. 30 %
  • Non-volatile matter:
    Max. 50 ppm
  • Residue on ignition:
    Max. 20 ppm
  • Free acid (as H₂SO₄):
    Max. 50 ppm
  • Silver (Ag):
    Max. 0.02 ppm
  • Aluminium (Al):
    Max. 0.2 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.02 ppm
  • Boron (B):
    Max. 0.02 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.02 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Gallium (Ga):
    Max. 0.02 ppm
  • Indium (In):
    Max. 0.02 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.02 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.2 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Platinum (Pt):
    Max. 0.02 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.02 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.02 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Chloride (Cl):
    Max. 0.5 ppm
  • Phosphate (PO₄):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 2 ppm
  • Total N:
    Max. 2 ppm

Specification Test Results

About this item

 :
Boiling Pt:Decomposes on heating
Boiling Pt: Dec. (on heating)
Melting Pt:-26 °C
Melting Pt:–26 °C
Density:1.11 g/cm³ (20 °C)
ADR:5.1,II
 :34.01 g/mol
EINECS:231-765-0
UN:2014
 :GHS07
 :GHS05
 :GHS03
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333