1-Butanol ≥99.4% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
Boiling Pt:119 °C (1.013 hPa)
Melting Pt:<–90 °C
Density:0.81 g/cm³ (20 °C)
Molecular Weight:74.12 g/mol
Flash Pt:9.4 %(V)
UN:1120
ADR:3 III
Merck Index:13,01539
Storage Temperature:Ambient
EINECS:200-751-6
REACH:01-2119484630-38
:GHS07
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Autoignition Temperature:649 °F
- Identity (IR):complying
- Assay (GC):Min. 99.40 %
- Non-volatile matter:Max. 0.005 %
- Water (Karl Fischer):Max. 0.1 %
- Titrable Acid:Max. 0.0008 meq/g
- Di-n-butyl ether (GC):Max. 0.2 %
- n-Butyraldehyde (GC):Max. 0.01 %
- APHA:Max. 10
- Explosion Limit:11.2 %
Specification Test Results
Frequently Bought Together