Order Entry
Poland
ContactUsLinkComponent
Organic reference standard, trans-2-Decenoyl-L-carnitine, analytical standard, Supelco®
Organic reference standard, trans-2-Decenoyl-L-carnitine, analytical standard, Supelco®
  56613-50MG
 :  Merck
 :  
Organic reference standard, trans-2-Decenoyl-L-carnitine, analytical standard, Supelco®
  56613-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • Description:
    trans-2-Decenoyl-L-carnitine, ≥94,0% (HPLC)
  • Seq:
    02
  • Formula:
    C₁₇H₃₁NO₄
  • Storage temperature:
    −20 °C
  • Description:
    trans-2-Decenoyl-L-carnitine