Order Entry
Poland
ContactUsLinkComponent
Organic reference standard, Potassium disulfite, analytical standard, Supelco®
Organic reference standard, Potassium disulfite, analytical standard, Supelco®
Catalog # 55777-1G
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Potassium disulfite, analytical standard, Supelco®
Catalog # 55777-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    16731-55-8
  • Description:
    Potassium disulfite, ≥98% (RT)
  • Formula:
     K₂S₂O₅
  • Description:
    Potassium disulfite
  • Form:
    Neat

Specifications

About this item