Order Entry
Poland
ContactUsLinkComponent
Organic reference standard, cis-3-Hexenyl acetate, analytical standard, Supelco®
Organic reference standard, cis-3-Hexenyl acetate, analytical standard, Supelco®
Catalog # 74597-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, cis-3-Hexenyl acetate, analytical standard, Supelco®
Catalog # 74597-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    3681-71-8
  • Description:
    cis-3-Hexenyl acetate, ≥98,0% (GC)
  • Formula:
    C₈H₁₄O₂  
  • Description:
    cis-3-Hexenyl acetate
  • Boiling point:
    75-76 °C/23 mm Hg(lit.)
  • Form:
    Neat

Specifications

About this item