Order Entry
Poland
ContactUsLinkComponent
NBD-PE membrane stain, for histology
NBD-PE membrane stain, for histology
Catalog # 60025.
Supplier:  Biotium
CAS Number:  
undefined
NBD-PE membrane stain, for histology
Catalog # 60025.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :956.24 g/mol
Storage Temperature:Freezer
CAS Number:178119-00-1
Formula:C₄₉H₉₀N₅O₁₁P