Tetraethylammonium hydrogen sulphate ≥99.0% for ion pair chromatography, Supelco®
Catalog # 86626-10G-F
Supplier: Merck
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:227.33 g/mol
CAS Number:16873-13-5
Formula:C₈H₂₁NO₄S
MDL Number:MFCD00036150