Order Entry
Poland
ContactUsLinkComponent
EPA 8270 GC-MS tuning solution, certified reference material, Supelco®
EPA 8270 GC-MS tuning solution, certified reference material, Supelco®
Catalog # 47387.
Supplier:  Merck
CAS Number:  
EPA 8270 GC-MS tuning solution, certified reference material, Supelco®
Catalog # 47387.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 mug/mL each component in dichloromethane
  • Pk:
    1
  • Description:
    EPA 8270 GC-MS Tuning Solution
  • Type:
    Certified reference material

Specifications

About this item