EPA 608 pesticides mix, certified reference material, Supelco®
Catalog # 47915-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:20 mug/mL each component in hexane: toluene (1:1)
- Pk:1
- Description:EPA 608 Pesticides Mix
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Type:Certified reference material