Supelco® Organic Reference Standard, Methyl Cis-15-Tetracosenoate
17265-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:2733-88-2
- Description:Methyl cis-15-tetracosenoate
- Storage conditions:–20 °C
- Shipping conditions:STD
- MDL No.:MFCD00056324
- M-Clarity quality level:MQ100
- Synonym(s):Nervonic acid methyl ester; Selacholeic acid methyl ester
- Formula:C₂₅H₄₈O₂
- Molecular weight:380,65
- Storage temperature:–20 °C
- Flash point:Not applicable
- GHS signal: GHS07