Supelco® PESTANAL® Organic Reference Standard, Carbofuran-3-Hydroxy
Catalog # 37896-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:16655-82-6
- Description:Carbofuran-3-hydroxy
- Environmentally Preferable:
- Storage conditions:–20 °C
- Hazard class description:H300
- Shipping conditions:STD
- MDL No.:MFCD00143168
- M-Clarity quality level:MQ100
- Melting point:140...148 °C
- Formula:C₁₂H₁₅NO₄
- Molecular weight:237,25
- Storage temperature:−20 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06
Caution: P264 - P270 - P301 + P310 - P405 - P501