Order Entry
Poland
ContactUsLinkComponent
Pharmaceutical analytical standard, Carbuterol-(tert-butyl-d9) acetate sesquihydrate, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Carbuterol-(tert-butyl-d9) acetate sesquihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1250-10MG
CAS Number:  
Pharmaceutical analytical standard, Carbuterol-(tert-butyl-d9) acetate sesquihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1250-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Carbuterol-(tert-butyl-d₉) acetate sesquihydrate
  • Formula:
    C13D9H12N3O3 · C2H4O2 · 1.5H2O
  • Storage temperature:
    2...8 °C
  • Description:
    Carbuterol-(tert-butyl-d₉) acetate sesquihydrate
  • Freezing point:
    110...115 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results