5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE], BioReagent suitable for fluorescence, Sigma-Aldrich®
Catalog # SIAL21955-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle, bottomless
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:527.52 g/mol
Storage Temperature:Freezer
CAS Number:150408-83-6
:246256-50-8
Formula:C₂₉H₂₅N₃O₇
MDL Number:MFCD00077324