
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥98.0% (by HPLC)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38894-11-0
Formula:C₈H₉N₃S·H₂O·HCl
MDL Number:MFCD00149370
:233.714 g/mol
Melting Pt:270...274 °C
EINECS:238-428-7
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Assay (HPLC):98.0%
Specification Test Results
Related Information
Frequently Bought Together