Order Entry
Poland
ContactUsLinkComponent
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) 10 mg/mL in water
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) 10 mg/mL in water
Catalog # 40044.
Supplier:  Biotium
CAS Number:  
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) 10 mg/mL in water
Catalog # 40044.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :533.88 g/mol
Storage Temperature:Refrigerator
EINECS:245-690-6
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl
MDL Number:MFCD00012679