Order Entry
Poland
ContactUsLinkComponent
Ammonium fluoride ≥98.0% eluent additive, for LC-MS, Fluka™
Ammonium fluoride ≥98.0% eluent additive, for LC-MS, Fluka™
Catalog # 52481-50G
CAS Number:  
Ammonium fluoride ≥98.0% eluent additive, for LC-MS, Fluka™
Catalog # 52481-50G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Ammonium hydrogen difluoride (NH₄HF₂):
    Max. 1 %
  • Residue on ignition (SO₄):
    Max. 0.005 %
  • Silver (Ag):
    Max. 5 ppm
  • Aluminium (Al):
    Max. 5 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 5 ppm
  • Thallium (Tl):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • Suitability for the LC-MS:
    complying

Specification Test Results

About this item

Synonyms: Fluoroammonium
 :
 :37.04 g/mol
EINECS:235-185-9
UN:2505
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:12125-01-8
Formula:NH₄F
MDL Number:MFCD00011423
Melting Pt:160
Density:1.01 g/cm³ (20 °C)
ADR:6.1 III
Merck Index:13,00524