Order Entry
Poland
ContactUsLinkComponent
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥98% (by TLC) amine-reactive fluorescent dye
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90022.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90022.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :527.52 g/mol
Storage Temperature:Freezer
CAS Number:150408-83-6
 :246256-50-8
Formula:C₂₉H₂₅N₃O₇
MDL Number:MFCD00077324